2,3-Dimethylbenzaldehyde ≥96.0%

Supplier: TCI America

D3463-1G D3463-5G
TCD3463-1GEA 34.76 CAD
TCD3463-1G TCD3463-5G
2,3-Dimethylbenzaldehyde ≥96.0%
2,3-Dimethylbenzaldehyde

CAS Number: 5779-93-1
MDL Number: MFCD00798004
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 102
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: o-Xylene-3-carboxaldehyde

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 110 °C
Density: 1.04 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00798004
CAS Number: 5779-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4a6163e262182ff7bb6ef1a88bdebd95f2ad57a&vid=a4a6163e262182ff7bb6ef1a88bdebd95f2ad57a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR