6-Hydroxy-3,4-dihydroquinolin-2(1H)-one ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 3,4-Dihydro-6-hydroxy-2(1H)-quinolinone, 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, Cilostazol Related Compound A, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline

D3448-5G D3448-25G
TCD3448-5GEA 84.75 CAD
TCD3448-5G TCD3448-25G
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one ≥98.0% (by HPLC, total nitrogen)
6-Hydroxy-3,4-dihydroquinolin-2(1H)-one

CAS Number: 54197-66-9
MDL Number: MFCD02179410
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 238


Other Product Names / Synonyms: 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline

Formula: C₉H₉NO₂
MW: 163.1700 g/mol
Melting Pt: 238 °C
MDL Number: MFCD02179410
CAS Number: 54197-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd87c65c030086cb71cb8f39692d6372f1b9d608&vid=cd87c65c030086cb71cb8f39692d6372f1b9d608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR