(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D3445-1G
D3445-100MG
TCD3445-1GEA
856.36
CAD
TCD3445-1G
TCD3445-100MG
(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)
(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine
CAS Number: 138517-66-5
MDL Number: MFCD06654768
Molecular Formula: C16H16N2
Molecular Weight: 236.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158
Specific rotation [a]20/D: 20 deg (C=1, MeOH)
Formula:
C₁₆H₁₆N₂ Melting Pt: 158 °C |
MDL Number:
MFCD06654768 CAS Number: 138517-66-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...