(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3444-100MG D3444-1G
TCD3444-100MGEA 97.49 CAD
TCD3444-100MG TCD3444-1G
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98.0% (by HPLC, titration analysis)
(11R,12R)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine

CAS Number: 181139-49-1
Molecular Formula: C16H16N2
Molecular Weight: 236.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 159
Specific rotation [a]20/D: -18 deg (C=1, H2O)

Formula: C₁₆H₁₆N₂
Melting Pt: 159 °C
MDL Number: MFCD09038520
CAS Number: 181139-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5ef02da394f625a2334d0b7b22a4adcedaec1d3&vid=b5ef02da394f625a2334d0b7b22a4adcedaec1d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR