2,4,6-Tris(3,4-dichlorophenyl)boroxin ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dichlorophenylboronic Anhydride, Tris(3,4-dichlorophenyl)cycloboroxane

D3435-5G D3435-25G
TCD3435-5GEA 206.66 CAD
TCD3435-5G TCD3435-25G
2,4,6-Tris(3,4-dichlorophenyl)boroxin ≥98.0%
2,4,6-Tris(3,4-dichlorophenyl)boroxin

CAS Number: 1133798-41-0
Molecular Formula: C18H9B3Cl6O3
Molecular Weight: 518.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Tris(3,4-dichlorophenyl)cycloboroxane, 3,4-Dichlorophenylboronic Anhydride

Formula: C₁₈H₉B₃Cl₆O₃
MW: 518.40 g/mol
CAS Number: 1133798-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa44e5d6c4486389439bd1e50e926569f97f84e5&vid=aa44e5d6c4486389439bd1e50e926569f97f84e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR