Dimethyl benzylmalonate ≥95.0%

Supplier: TCI America

D3432-5G D3432-25G
TCD3432-5GEA 123.24 CAD
TCD3432-5G TCD3432-25G
Dimethyl benzylmalonate ≥95.0%
Dimethyl benzylmalonate

CAS Number: 49769-78-0
MDL Number: MFCD01445471
Molecular Formula: C12H14O4
Molecular Weight: 222.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 285
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Benzylmalonic Acid Dimethyl Ester

Formula: C₁₂H₁₄O₄
Boiling Pt: 285 °C
Density: 1.13 (20 °C)
MDL Number: MFCD01445471
CAS Number: 49769-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75a1e57162f1a601f688530ce42b5a88128e4377&vid=75a1e57162f1a601f688530ce42b5a88128e4377&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR