1,3-Difluoro-5-nitrobenzene ≥98.0%

Supplier: TCI America

D3426-25G D3426-5G
TCD3426-25GEA 586.81 CAD
TCD3426-25G TCD3426-5G
1,3-Difluoro-5-nitrobenzene ≥98.0%
1,3-Difluoro-5-nitrobenzene

CAS Number: 2265-94-3
MDL Number: MFCD00012142
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 73
Freezing point (°C): 18
Specific Gravity (20/20): 1.43


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3-Difluoro-5-nitrobenzene

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 177 °C
Density: 1.43 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00012142
CAS Number: 2265-94-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abd2a30fbfb78e387800b080b2b3f54f14674fb2&vid=36abb61c4f2b64c1bd77241a6448001da3ea5ad2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR