DL-1,4-Dichloro-2,3-butanediol ≥98.0%
Supplier: TCI America
D3423-5G
D3423-25G
TCD3423-5GEA
252.96
CAD
TCD3423-5G
TCD3423-25G
DL-1,4-Dichloro-2,3-butanediol ≥98.0%
DL-1,4-Dichloro-2,3-butanediol
CAS Number: 2419-73-0
Molecular Formula: C4H8Cl2O2
Molecular Weight: 159.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 152
Melting point (°C): 77
Formula:
C₄H₈Cl₂O₂ MW: 159.01 g/mol Boiling Pt: 152 °C Melting Pt: 77 °C |
CAS Number:
2419-73-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...