2,2-Dimethyl-1,3-benzodioxole ≥98.0%
Supplier: TCI America
Synonyms:
1,2-(Isopropylidenedioxy)benzene, 2,2-Dimethylbenzo[d][1,3]dioxole
D3421-5G
D3421-25G
TCD3421-5GEA
170.64
CAD
TCD3421-5G
TCD3421-25G
2,2-Dimethyl-1,3-benzodioxole ≥98.0%
2,2-Dimethyl-1,3-benzodioxole
CAS Number: 14005-14-2
MDL Number: MFCD08061594
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Melting point (°C): 3
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1,2-(Isopropylidenedioxy)benzene
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 182 °C Melting Pt: 3 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD08061594 CAS Number: 14005-14-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...