Diisobutyl phosphonate ≥95.0%

Supplier: TCI America

D3417-500G D3417-25G
TCD3417-500GEA 229.1 CAD
TCD3417-500G TCD3417-25G
Diisobutyl phosphonate ≥95.0%
Diisobutyl phosphonate

CAS Number: 1189-24-8
Molecular Formula: C8H19O3P
Molecular Weight: 194.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, Dialkyl Phosphites (Multi-component Reaction)
Other Product Names / Synonyms: Phosphorous Acid Diisobutyl Ester, Diisobutyl Phosphonate, Phosphonic Acid Diisobutyl Ester

Formula: C₈H₁₉O₃P
Boiling Pt: 138 °C
Density: 0.98 (20 °C)
CAS Number: 1189-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d97792af7ba5de6da6f078bedbdea9bf38323ab4&vid=d97792af7ba5de6da6f078bedbdea9bf38323ab4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR