Diethylene glycol monobenzyl ether ≥98.0%

Supplier: TCI America

Synonyms: 2-[2-(Benzyloxy)ethoxy]ethanol, 2-(2-(Benzyloxy)ethoxy)ethanol

D3408-25G D3408-500G
TCD3408-25GEA 79 CAD
TCD3408-25G TCD3408-500G
Diethylene glycol monobenzyl ether ≥98.0%
Diethylene glycol monobenzyl ether

CAS Number: 2050-25-1
MDL Number: MFCD00029649
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 158
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 2-[2-(Benzyloxy)ethoxy]ethanol

Formula: C₁₁H₁₆O₃
Density: 1.08 (20 °C)
Flash Pt: 158 °C
MDL Number: MFCD00029649
CAS Number: 2050-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b31a8351fcf28d5e6041ecde5e62c20c5d4eee73&vid=b31a8351fcf28d5e6041ecde5e62c20c5d4eee73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR