Dicyclopentanyl methacrylate ≥95.0% stabilized

Supplier: TCI America

D3381-500G D3381-25G D3381-100G
TCD3381-500GEA 368.14 CAD
TCD3381-500G TCD3381-25G TCD3381-100G
Dicyclopentanyl methacrylate ≥95.0% stabilized
Dicyclopentanyl methacrylate

(stabilized with MEHQ)
CAS Number: 34759-34-7
Molecular Formula: C14H20O2
Molecular Weight: 220.31
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 145
Specific Gravity (20/20): 1.04
Storage Temperature: <0°C


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: Methacrylic Acid Dicyclopentanyl Ester, Methacrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Methacrylate

Formula: C₁₄H₂₀O₂
Density: 1.04 (20 °C)
Flash Pt: 145 °C
MDL Number: MFCD28963492
CAS Number: 34759-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d7397e85c5047d535dad29545f5a59a16add0a&vid=05d7397e85c5047d535dad29545f5a59a16add0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR