Dicyclopentanyl acrylate ≥95.0% stabilized
Supplier: TCI America
D3380-25G
D3380-500G
TCD3380-25GEA
56.88
CAD
TCD3380-25G
TCD3380-500G
Dicyclopentanyl acrylate ≥95.0% stabilized
Dicyclopentanyl acrylate
(stabilized with MEHQ)
CAS Number: 7398-56-3
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 132
Specific Gravity (20/20): 1.06
Storage Temperature: <0°C
Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: Acrylic Acid Dicyclopentanyl Ester, Acrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Acrylate
Formula:
C₁₃H₁₈O₂ Density: 1.06 (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD24663518 CAS Number: 7398-56-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...