N,4'-Dimethylformanilide ≥97.0%
Supplier: TCI America
D3369-1G
D3369-5G
TCD3369-1GEA
418.7
CAD
TCD3369-1G
TCD3369-5G
N,4'-Dimethylformanilide ≥97.0%
N,4'-Dimethylformanilide
CAS Number: 2739-04-0
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 277
Other Product Names / Synonyms: N-Methyl-N-(p-tolyl)formamide
Formula:
C₉H₁₁NO Boiling Pt: 277 °C |
CAS Number:
2739-04-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...