N,2'-Dimethylformanilide ≥97.0%
Supplier: TCI America
Synonyms:
N-Methyl-N-(o-tolyl)formamide
D3368-1G
D3368-5G
TCD3368-1GEA
190.07
CAD
TCD3368-1G
TCD3368-5G
N,2'-Dimethylformanilide ≥97.0%
N,2'-Dimethylformanilide
CAS Number: 131840-54-5
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Other Product Names / Synonyms: N-Methyl-N-(o-tolyl)formamide
Formula:
C₉H₁₁NO MW: 149.19 g/mol Boiling Pt: 206 °C |
MDL Number:
MFCD09038514 CAS Number: 131840-54-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...