3,5-Dimethyl-1-adamantanemethanol ≥98.0%

Supplier: TCI America

Synonyms: 1-(Hydroxymethyl)-3,5-dimethyladamantane

D3367-1G D3367-5G
TCD3367-1GEA 104.28 CAD
TCD3367-1G TCD3367-5G
3,5-Dimethyl-1-adamantanemethanol ≥98.0%
3,5-Dimethyl-1-adamantanemethanol

CAS Number: 26919-42-6
Molecular Formula: C13H22O
Molecular Weight: 194.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 60


Other Product Names / Synonyms: 1-(Hydroxymethyl)-3,5-dimethyladamantane

Formula: C₁₃H₂₂O
MW: 194.32 g/mol
Boiling Pt: 115 °C
Melting Pt: 60 °C
MDL Number: MFCD00188062
CAS Number: 26919-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=283f6bddf9999b5180b615c3e04381419d940740&vid=283f6bddf9999b5180b615c3e04381419d940740&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR