2,6-Dichlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2,6-Dichlorophenyl)metaboric acid, 2,6-Dichlorobenzeneboronic acid, Dihydroxy(2,6-dichlorophenyl)borane, (2,6-Dichlorophenyl)boronic acid

D3357-5G D3357-1G
TCD3357-5GEA 211.72 CAD
TCD3357-5G 76219-686
2,6-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
2,6-Dichlorophenylboronic acid

CAS Number: 73852-17-2
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: White
Melting point (°C): 151


Other Product Names / Synonyms: 2,6-Dichlorobenzeneboronic Acid

Formula: C₆H₅BCl₂O₂
MW: 190.82
Melting Pt: 151 °C
MDL Number: MFCD00064869
CAS Number: 73852-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e5cfe41086bd9db1e02891fc5a12ee35cd0470&vid=24e5cfe41086bd9db1e02891fc5a12ee35cd0470&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR