Diethyl (2-oxo-2-phenylethyl)phosphonate ≥97.0%

Supplier: TCI America

Synonyms: Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester

D3339-1G D3339-5G
TCD3339-1GEA 233.84 CAD
TCD3339-1G TCD3339-5G
Diethyl (2-oxo-2-phenylethyl)phosphonate ≥97.0%
Diethyl (2-oxo-2-phenylethyl)phosphonate

CAS Number: 3453-00-7
Molecular Formula: C12H17O4P
Molecular Weight: 256.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 113
Specific Gravity (20/20): 1.18


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester

Formula: C₁₂H₁₇O₄P
Boiling Pt: 193 °C
Density: 1.18 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00015342
CAS Number: 3453-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10a36c66bbe5cf6050b9c73719d972047341d042&vid=10a36c66bbe5cf6050b9c73719d972047341d042&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR