5-Amino-1-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Phenyl-1H-pyrazol-5-amine, 5-Amino-1-phenylpyrazole

A0174-5G A0174-25G
TCA0174-005GEA 308.1 CAD
TCA0174-005G TCA0174-25G
5-Amino-1-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
5-Amino-1-phenyl-1H-pyrazole

CAS Number: 826-85-7
MDL Number: MFCD00020734
Molecular Formula: C9H9N3
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 44

Formula: C₉H₉N₃
MW: 159.1900 g/mol
Melting Pt: 44 °C
MDL Number: MFCD00020734
CAS Number: 826-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f575f7a004a27e774f31c13e1032b306427063&vid=38f575f7a004a27e774f31c13e1032b306427063&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR