Diethyl (3-methylbenzyl)phosphonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 3-methylbenzylphosphonate

D3328-25G D3328-5G
TCD3328-25GEA 148.84 CAD
TCD3328-25G TCD3328-5G
Diethyl (3-methylbenzyl)phosphonate ≥98.0%
Diethyl (3-methylbenzyl)phosphonate

CAS Number: 63909-50-2
Molecular Formula: C12H19O3P
Molecular Weight: 242.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.08


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 3-(Diethylphosphonomethyl)toluene, (3-Methylbenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₂H₁₉O₃P
MW: 242.2512 g/mol
Boiling Pt: 127 °C
Density: 1.08 (20 °C)
MDL Number: MFCD08276364
CAS Number: 63909-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c994d5798560acfd0c95f19e35299248199a4416&vid=c994d5798560acfd0c95f19e35299248199a4416&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR