Diethyl (2-methylbenzyl)phosphonate ≥98.0%
Supplier: TCI America
D3327-5G
D3327-25G
TCD3327-5GEA
118.97
CAD
TCD3327-5G
TCD3327-25G
Diethyl (2-methylbenzyl)phosphonate ≥98.0%
Diethyl (2-methylbenzyl)phosphonate
CAS Number: 62778-16-9
Molecular Formula: C12H19O3P
Molecular Weight: 242.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.09
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 2-(Diethylphosphonomethyl)toluene, (2-Methylbenzyl)phosphonic Acid Diethyl Ester
Formula:
C₁₂H₁₉O₃P Density: 1.09 (20 °C) |
CAS Number:
62778-16-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...