Diethyl (3-methoxybenzyl)phosphonate ≥98.0%

Supplier: TCI America

D3326-5G D3326-25G
TCD3326-5GEA 140.46 CAD
TCD3326-5G TCD3326-25G
Diethyl (3-methoxybenzyl)phosphonate ≥98.0%
Diethyl (3-methoxybenzyl)phosphonate

CAS Number: 60815-18-1
Molecular Formula: C12H19O4P
Molecular Weight: 258.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.13


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 3-(Diethylphosphonomethyl)anisole, (3-Methoxybenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₂H₁₉O₄P
Boiling Pt: 135 °C
Density: 1.13 (20 °C)
MDL Number: MFCD08276363
CAS Number: 60815-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72631f7f6ca335e1e070386de7dcb2e7099e1da&vid=e72631f7f6ca335e1e070386de7dcb2e7099e1da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR