Diethyl (4-fluorobenzyl)phosphonate ≥98.0%
Supplier: TCI America
Synonyms:
(4-Fluorobenzyl)phosphonic acid diethyl ester, Diethyl 4-fluorobenzylphosphonate
D3324-5G
D3324-25G
TCD3324-5GEA
66.36
CAD
TCD3324-5G
TCD3324-25G
Diethyl (4-fluorobenzyl)phosphonate ≥98.0%
Diethyl (4-fluorobenzyl)phosphonate
CAS Number: 63909-58-0
Molecular Formula: C11H16FO3P
Molecular Weight: 246.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Specific Gravity (20/20): 1.16
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Fluorinated Building Blocks
Other Product Names / Synonyms: (4-Fluorobenzyl)phosphonic Acid Diethyl Ester
Formula:
C₁₁H₁₆FO₃P MW: 246.2200 g/mol Boiling Pt: 150 °C Density: 1.16 (20 °C) |
MDL Number:
MFCD06795966 CAS Number: 63909-58-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...