3,4-Dihydro-2H-pyran-2-methanol ≥97.0%

Supplier: TCI America

Synonyms: 2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker

D3295-1G D3295-5G
TCD3295-1GEA 44.24 CAD
TCD3295-1G TCD3295-5G
3,4-Dihydro-2H-pyran-2-methanol ≥97.0%
3,4-Dihydro-2H-pyran-2-methanol

CAS Number: 3749-36-8
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 85
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 2-Hydroxymethyl-3,4-dihydro-2H-pyran

Formula: C₆H₁₀O₂
Boiling Pt: 93 °C
Density: 1.07 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00051507
CAS Number: 3749-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b362e9c2ecbea02c04a89677ec8a01c8fca5dbb&vid=6b362e9c2ecbea02c04a89677ec8a01c8fca5dbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR