3,4-Dichloro-6-nitrotoluene ≥98.0%

Supplier: TCI America

Synonyms: 4,5-Dichloro-2-nitrotoluene, 1,2-Dichloro-4-methyl-5-nitrobenzene

D3271-25G D3271-5G
TCD3271-25GEA 314.42 CAD
TCD3271-25G TCD3271-5G
3,4-Dichloro-6-nitrotoluene ≥98.0%
3,4-Dichloro-6-nitrotoluene

CAS Number: 7494-45-3
Molecular Formula: C7H5Cl2NO2
Molecular Weight: 206.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 62 °C
MDL Number: MFCD01109847
CAS Number: 7494-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42f93a2f4036e905dddfa169d95eede0ab04de6e&vid=42f93a2f4036e905dddfa169d95eede0ab04de6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR