6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
D3265-25G
D3265-100G
TCD3265-25GEA
71.04
CAD
TCD3265-25G
TCD3265-100G
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol ≥98.0% (by HPLC, total nitrogen)
6-(Dibutylamino)-1,3,5-triazine-2,4-dithiol
CAS Number: 29529-99-5
Molecular Formula: C11H20N4S2
Molecular Weight: 272.43
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 143
Other Product Names / Synonyms: 2-(Dibutylamino)-4,6-dimercapto-1,3,5-triazine
Formula:
C₁₁H₂₀N₄S₂ Melting Pt: 143 °C |
MDL Number:
MFCD00272594 CAS Number: 29529-99-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...