Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl ≥97.0%

Supplier: TCI America

Synonyms: Sodium 2,2'-([1,1'-biphenyl]-4,4'-diylbis(ethene-2,1-diyl))dibenzenesulfonate, 4,4'-Bis(2-sulfonatostyryl)biphenyl Disodium Salt, Sodium 2,2'-([1,1'-biphenyl]-4,4'-diylbis(ethene-2,1-diyl))dibenzenesulphonate, 4,4'-Bis(2-sulphonatostyryl)biphenyl Disodium Salt

D3251-25G D3251-100G
TCD3251-25GEA 121.66 CAD
TCD3251-25G TCD3251-100G
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl ≥97.0%
Disodium-4,4'-bis(2-sulfonatostyryl)biphenyl

CAS Number: 27344-41-8
Molecular Formula: C28H22O6S2
Molecular Weight: 562.56
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Fluorescent Stains
Other Product Names / Synonyms: 4,4'-Bis(2-sulfonatostyryl)biphenyl Disodium Salt

Formula: C₂₈H₂₂O₆S₂
MW: 562.56 g/mol
MDL Number: MFCD00318874
CAS Number: 27344-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80e351dcb0f2ba3fa4f529681df3b795f2573fec&vid=80e351dcb0f2ba3fa4f529681df3b795f2573fec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR