9,9-Dimethylfluorene ≥98.0%

Supplier: TCI America

D3235-5G D3235-1G
TCD3235-5GEA 311.26 CAD
TCD3235-5G TCD3235-1G
9,9-Dimethylfluorene ≥98.0%
9,9-Dimethylfluorene

CAS Number: 4569-45-3
MDL Number: MFCD00114670
Molecular Formula: C15H14
Molecular Weight: 194.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 287
Melting point (°C): 97
Lambda max.: 301 nm (EtOH)

Formula: C₁₅H₁₄
Boiling Pt: 287 °C
Melting Pt: 97 °C
MDL Number: MFCD00114670
CAS Number: 4569-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfc75e928257bcf10478481dc4272d59cd265868&vid=bfc75e928257bcf10478481dc4272d59cd265868&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR