2,4-Diethylglutaric acid (DL- and meso- mixture) ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Diethylpentanedioic Acid

D3234-5G D3234-25G
TCD3234-5GEA 363.4 CAD
TCD3234-5G TCD3234-25G
2,4-Diethylglutaric acid (DL- and meso- mixture) ≥98.0%
2,4-Diethylglutaric acid (DL- and meso- mixture)

CAS Number: 27899-21-4
MDL Number: MFCD04117827
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 78


Other Product Names / Synonyms: 2,4-Diethylpentanedioic Acid

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Melting Pt: 78 °C
MDL Number: MFCD04117827
CAS Number: 27899-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20c3fcbd4c2742b8952423b9c9071d2a9e0849f9&vid=20c3fcbd4c2742b8952423b9c9071d2a9e0849f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR