2-(Diethylamino)ethyl bromide hydrobromide ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromotriethylamine Hydrobromide, 2-Bromoethyldiethylamine Hydrobromide, 2-Bromo-N,N-diethylethylamine Hydrobromide

D3232-25G D3232-500G D3232-100G
TCD3232-25GEA 200.66 CAD
TCD3232-25G TCD3232-500G TCD3232-100G
2-(Diethylamino)ethyl bromide hydrobromide ≥98.0%
2-(Diethylamino)ethyl bromide hydrobromide

CAS Number: 1069-72-3
Molecular Formula: C6H14BrN
Molecular Weight: 261.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 208


Other Product Names / Synonyms: 2-Bromo-N,N-diethylethylamine Hydrobromide, 2-Bromoethyldiethylamine Hydrobromide, 2-Bromotriethylamine Hydrobromide

Formula: C₆H₁₄BrN
MW: 261.00 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00040376
CAS Number: 1069-72-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ece4e28f8b7cc80887957140de3001fff230234d&vid=ece4e28f8b7cc80887957140de3001fff230234d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR