2,4-Dimethyl-5-phenylthiophene ≥98.0%

Supplier: TCI America

D3228-1G D3228-5G
TCD3228-1GEA 127.35 CAD
TCD3228-1G TCD3228-5G
2,4-Dimethyl-5-phenylthiophene ≥98.0%
2,4-Dimethyl-5-phenylthiophene

CAS Number: 57021-49-5
Molecular Formula: C12H12S
Molecular Weight: 188.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C

Formula: C₁₂H₁₂S
MW: 188.29 g/mol
Boiling Pt: 151 °C
Density: 1.08 (20 °C)
CAS Number: 57021-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da35aee0127e2587e258e43c88c5e08e0016af0f&vid=da35aee0127e2587e258e43c88c5e08e0016af0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR