trans-N,N'-Diacetylcyclohexane-1,2-diamine ≥98.0%

Supplier: TCI America

D3226-5G D3226-25G
TCD3226-5GEA 162.74 CAD
TCD3226-5G TCD3226-25G
trans-N,N'-Diacetylcyclohexane-1,2-diamine ≥98.0%
trans-N,N'-Diacetylcyclohexane-1,2-diamine

CAS Number: 70924-78-6
Molecular Formula: C10H18N2O2
Molecular Weight: 198.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 270


Other Product Names / Synonyms: trans-1,2-Bis(acetamido)cyclohexane

Formula: C₁₀H₁₈N₂O₂
Melting Pt: 270 °C
MDL Number: MFCD08276353
CAS Number: 70924-78-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c859c3e66ab372ddd579a89b5c314a6c59f8895&vid=5c859c3e66ab372ddd579a89b5c314a6c59f8895&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR