4-(Dimethylamino)pyridine-N-oxide hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D3220-5G D3220-1G
TCD3220-5GEA 922.72 CAD
TCD3220-5G TCD3220-1G
4-(Dimethylamino)pyridine-N-oxide hydrate ≥98.0% (by titrimetric analysis)
4-(Dimethylamino)pyridine-N-oxide hydrate

CAS Number: 1005-31-8
Molecular Formula: C7H10N2O
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 97


Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: DMAPO

Formula: C₇H₁₀N₂O
Melting Pt: 97 °C
CAS Number: 1005-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b151ae5c36132cfe08e1a925b56b8b567baf253&vid=4b151ae5c36132cfe08e1a925b56b8b567baf253&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR