2,3-Dimethyl-2,3-butanediamine dihydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
2,3-Diamino-2,3-dimethylbutane Dihydrochloride, 1,1,2,2-Tetramethylethylenediamine Dihydrochloride
D3212-1G
D3212-5G
TCD3212-1GEA
203.82
CAD
TCD3212-1G
TCD3212-5G
2,3-Dimethyl-2,3-butanediamine dihydrochloride ≥98.0%
2,3-Dimethyl-2,3-butanediamine dihydrochloride
CAS Number: 75804-28-3
Molecular Formula: C6H16N2
Molecular Weight: 189.12
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3-Diamino-2,3-dimethylbutane Dihydrochloride, 1,1,2,2-Tetramethylethylenediamine Dihydrochloride
Formula:
C₆H₁₆N₂ MW: 189.12 g/mol |
CAS Number:
75804-28-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...