3,6-Di(4-pyridyl)-1,2,4,5-tetrazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3211-1G D3211-5G
TCD3211-1GEA 240.16 CAD
TCD3211-1G TCD3211-5G
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine ≥98.0% (by HPLC, titration analysis)
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine

CAS Number: 57654-36-1
Molecular Formula: C12H8N6
Molecular Weight: 236.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 257


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 3,6-Bis(4-pyridyl)-1,2,4,5-tetrazine

Formula: C₁₂H₈N₆
Melting Pt: 257 °C
MDL Number: MFCD00129054
CAS Number: 57654-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff476e2e7b719c8e73a5826abd528f061bfa1d67&vid=ff476e2e7b719c8e73a5826abd528f061bfa1d67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR