Diammonium ethylenediaminetetraacetate monohydrate ≥98.0%

Supplier: TCI America

Synonyms: Diammonium EDTA, Ammonium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate hydrate, Ethylenediaminetetraacetic Acid Diammonium Salt

D3195-25G D3195-500G
TCD3195-25GEA 61.07 CAD
TCD3195-25G TCD3195-500G
Diammonium ethylenediaminetetraacetate monohydrate ≥98.0%
Diammonium ethylenediaminetetraacetate monohydrate

CAS Number: 304675-80-7 Molecular Formula: C10H16N2O8 Molecular Weight: 326.31 Purity/Analysis Method: 98.0% (N,T) Form: Crystal Melting point (°C): 218


Other Product Names / Synonyms: Diammonium EDTA, Ethylenediaminetetraacetic Acid Diammonium Salt

Formula: C₁₀H₂₄N₄O₉
MW: 344.32 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00150463
CAS Number: 304675-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ae293fd4234c56c5549b406dfec447cc143b0e9&vid=8ae293fd4234c56c5549b406dfec447cc143b0e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR