3,5-Bis(benzyloxy)benzaldehyde ≥98.0%

Supplier: TCI America

D3184-1G D3184-5G
TCD3184-1GEA 69.36 CAD
TCD3184-1G TCD3184-5G
3,5-Bis(benzyloxy)benzaldehyde ≥98.0%
3,5-Bis(benzyloxy)benzaldehyde

CAS Number: 14615-72-6
Molecular Formula: C21H18O3
Molecular Weight: 318.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(benzyloxy)benzaldehyde

Formula: C₂₁H₁₈O₃
Melting Pt: 82 °C
MDL Number: MFCD00075777
CAS Number: 14615-72-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fdde93daa9d83d07a071f28263d3dc0b9c921b4&vid=8fdde93daa9d83d07a071f28263d3dc0b9c921b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR