2,6-Dibromo-9,10-anthraquinone ≥95.0%

Supplier: TCI America

Synonyms: 2,6-Dibromoanthracene-9,10-dione

D3182-5G D3182-1G
TCD3182-5GEA 925.56 CAD
TCD3182-5G TCD3182-1G
2,6-Dibromo-9,10-anthraquinone ≥95.0%
2,6-Dibromo-9,10-anthraquinone

CAS Number: 633-70-5
Molecular Formula: C14H6Br2O2
Molecular Weight: 366.01
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 290

Formula: C₁₄H₆Br₂O₂
MW: 366.0042 g/mol
Melting Pt: 290 °C
MDL Number: MFCD08276344
CAS Number: 633-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba9e74dce6a1fe978b1865c2f647d14fb2aeb855&vid=ba9e74dce6a1fe978b1865c2f647d14fb2aeb855&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR