4-(1,1-Dioxothiomorpholino)benzaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
4-(1,1-Dioxidothiomorpholino)benzaldehyde, 4-(4-Formylphenyl)thiomorpholine 1,1-Dioxide
D3162-5G
D3162-1G
TCD3162-5GEA
583.49
CAD
TCD3162-5G
TCD3162-1G
4-(1,1-Dioxothiomorpholino)benzaldehyde ≥98.0%
4-(1,1-Dioxothiomorpholino)benzaldehyde
CAS Number: 27913-96-8
Molecular Formula: C11H13NO3S
Molecular Weight: 239.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 185
Other Product Names / Synonyms: 4-(4-Formylphenyl)thiomorpholine 1,1-Dioxide
Formula:
C₁₁H₁₃NO₃S MW: 239.29 g/mol Melting Pt: 185 °C |
MDL Number:
MFCD08276340 CAS Number: 27913-96-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...