3,3-Dimethyl-1-indanone ≥95.0%

Supplier: TCI America

D3160-5G D3160-25G D3160-1G
TCD3160-5GEA 376.04 CAD
TCD3160-5G TCD3160-25G TCD3160-1G
3,3-Dimethyl-1-indanone ≥95.0%
3,3-Dimethyl-1-indanone

CAS Number: 26465-81-6
MDL Number: MFCD01846169
Molecular Formula: C11H12O
Molecular Weight: 160.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C

Formula: C₁₁H₁₂O
Boiling Pt: 122 °C
Density: 1.03 (20 °C)
MDL Number: MFCD01846169
CAS Number: 26465-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=683aac3974c311729fd509476d346f9d62ac3354&vid=683aac3974c311729fd509476d346f9d62ac3354&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR