4,4'-Dimethoxy-trans-stilbene ≥98.0%
Supplier: TCI America
D3104-500MG
D3104-1G
TCD3104-500MGEA
260.7
CAD
TCD3104-500MG
TCD3104-1G
4,4'-Dimethoxy-trans-stilbene ≥98.0%
4,4'-Dimethoxy-trans-stilbene
CAS Number: 15638-14-9
Molecular Formula: C16H16O2
Molecular Weight: 240.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 214
Storage Temperature: 0-10°C
Formula:
C₁₆H₁₆O₂ Melting Pt: 214 °C |
CAS Number:
15638-14-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...