4,4'-Diiodo-trans-stilbene ≥97.0%
Supplier: TCI America
D3094-1G
D3094-5G
TCD3094-1GEA
255.96
CAD
TCD3094-1G
TCD3094-5G
4,4'-Diiodo-trans-stilbene ≥97.0%
4,4'-Diiodo-trans-stilbene
CAS Number: 201861-91-8
Molecular Formula: C14H10I2
Molecular Weight: 432.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Formula:
C₁₄H₁₀I₂ |
CAS Number:
201861-91-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...