4,4'-Dichloro-trans-stilbene ≥98.0%
Supplier: TCI America
D3063-1G
D3063-250MG
TCD3063-1GEA
733.91
CAD
TCD3063-1G
TCD3063-250MG
4,4'-Dichloro-trans-stilbene ≥98.0%
4,4'-Dichloro-trans-stilbene
CAS Number: 1657-56-3
Molecular Formula: C14H10Cl2
Molecular Weight: 249.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 176
Storage Temperature: 0-10°C
Formula:
C₁₄H₁₀Cl₂ Boiling Pt: 230 °C Melting Pt: 176 °C |
MDL Number:
MFCD01108802 CAS Number: 1657-56-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...