4,4'-Dibromo-trans-stilbene ≥98.0%
Supplier: TCI America
Synonyms:
(E)-1,2-Bis(4-bromophenyl)ethene
D3043-1G
D3043-5G
D3043-250MG
TCD3043-1GEA
77.42
CAD
TCD3043-1G
TCD3043-5G
TCD3043-250MG
4,4'-Dibromo-trans-stilbene ≥98.0%
4,4'-Dibromo-trans-stilbene
CAS Number: 18869-30-2
Molecular Formula: C14H10Br2
Molecular Weight: 338.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 212
Formula:
C₁₄H₁₀Br₂ MW: 338.0372 g/mol Melting Pt: 212 °C |
MDL Number:
MFCD00448009 CAS Number: 18869-30-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...