1,2'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI America
D2988-1G
D2988-5G
TCD2988-1GEA
192.76
CAD
TCD2988-1G
TCD2988-5G
1,2'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
1,2'-Dinaphthylamine
CAS Number: 4669-06-1
Molecular Formula: C20H15N
Molecular Weight: 269.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 111
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Highly Purified Reagents
Other Product Names / Synonyms: 1,2'-Iminodinaphthalene
Formula:
C₂₀H₁₅N Melting Pt: 111 °C |
MDL Number:
MFCD01632468 CAS Number: 4669-06-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...