2,2'-Dinaphthylamine ≥98.0%

Supplier: TCI America

D2987-1G D2987-5G
TCD2987-1GEA 131.14 CAD
TCD2987-1G TCD2987-5G
2,2'-Dinaphthylamine ≥98.0%
2,2'-Dinaphthylamine

CAS Number: 532-18-3
MDL Number: MFCD03093264
Molecular Formula: C20H15N
Molecular Weight: 269.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 471
Melting point (°C): 174


Other Product Names / Synonyms: 2,2'-Iminodinaphthalene

Formula: C₂₀H₁₅N
Boiling Pt: 471 °C
Melting Pt: 174 °C
MDL Number: MFCD03093264
CAS Number: 532-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81bd550ce4984cf1e688cad3529a42d0e7f8ffb9&vid=81bd550ce4984cf1e688cad3529a42d0e7f8ffb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR