3,6-Dibromo-9-ethylcarbazole ≥98.0%

Supplier: TCI America

Synonyms: 3,6-Dibromo-9-ethyl-9H-carbazole

D2982-5G D2982-1G
TCD2982-5GEA 114.08 CAD
TCD2982-5G TCD2982-1G
3,6-Dibromo-9-ethylcarbazole ≥98.0%
3,6-Dibromo-9-ethylcarbazole

CAS Number: 33255-13-9
Molecular Formula: C14H11Br2N
Molecular Weight: 353.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 141

Formula: C₁₄H₁₁Br₂N
MW: 353.06 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00138134
CAS Number: 33255-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d871073ca4450e5087b7069e7658e2b0af223087&vid=d871073ca4450e5087b7069e7658e2b0af223087&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR