3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
3,6-Dibromo-9-phenyl-9H-carbazole
D2981-1G
D2981-5G
TCD2981-1GEA
66.36
CAD
TCD2981-1G
TCD2981-5G
3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
3,6-Dibromo-9-phenylcarbazole
CAS Number: 57103-20-5
Molecular Formula: C18H11Br2N
Molecular Weight: 401.10
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Formula:
C₁₈H₁₁Br₂N MW: 401.10 g/mol |
MDL Number:
MFCD08276338 CAS Number: 57103-20-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...