2,3-Dimethylanthraquinone ≥95.0%
Supplier: TCI America
D2980-1G
D2980-5G
TCD2980-1GEA
197.5
CAD
TCD2980-1G
TCD2980-5G
2,3-Dimethylanthraquinone ≥95.0%
2,3-Dimethylanthraquinone
CAS Number: 6531-35-7
Molecular Formula: C16H12O2
Molecular Weight: 236.27
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 210
Formula:
C₁₆H₁₂O₂ Melting Pt: 210 °C |
CAS Number:
6531-35-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...