1,3,2-Dioxathiolane-2-oxide ≥98.0%

Supplier: TCI America

D2977-25G D2977-5G
TCD2977-25GEA 61.62 CAD
TCD2977-25G TCD2977-5G
1,3,2-Dioxathiolane-2-oxide ≥98.0%
1,3,2-Dioxathiolane-2-oxide

CAS Number: 3741-38-6
Molecular Formula: C2H4O3S
Molecular Weight: 108.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 114
Specific Gravity (20/20): 1.44


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Ethylene Sulfite

Formula: C₂H₄O₃S
Boiling Pt: 172 °C
Density: 1.44 (20 °C)
Flash Pt: 114 °C
MDL Number: MFCD00005354
CAS Number: 3741-38-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fcaf444b15b021a7989fe604e18e9da48d2b85f&vid=6fcaf444b15b021a7989fe604e18e9da48d2b85f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR